| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10020304.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10020304.hg.1 |
ENST00000535447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10020304.hg.1 |
ENST00000543866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10020304.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10020304.hg.1 |
TC10001580.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000178977.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178977.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178977.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000178977.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178977.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178977.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000178977.hg.1 |
TC1000011585.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000178979.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178979.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178979.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000178979.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178979.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178979.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000178979.hg.1 |
TC1000011585.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000370508.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000471741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
ENST00000489349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011585.hg.1 |
NM_002079.2 |
Refseq |