| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01012428.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01012428.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01012428.hg.1 |
TC01000778.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100162558.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100162558.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100162558.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100162558.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100162558.hg.1 |
TC0100008773.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
ENST00000263187.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008773.hg.1 |
NM_002440.3 |
Refseq |