Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01549373_m1
          See other SEMA5A GE Assays ›
          Gene Symbol:
          SEMA5A
          Gene Name:
          semaphorin 5A
          Gene Aliases:
          SEMAF, semF
          Chromosome Location:
          Chr.5: 9035026 - 9546121 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01549373_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SEMA5A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003966.2 NP_003957.2 15 - 16 2640 80
          XM_006714506.2 XP_006714569.1 16 - 17 2739 80
          XM_006714507.3 XP_006714570.1 14 - 15 2302 80
          XM_011514155.2 XP_011512457.1 15 - 16 2263 80
          XM_011514156.1 XP_011512458.1 17 - 18 2812 80
          XM_011514157.1 XP_011512459.1 16 - 17 2667 80
          XM_011514158.1 XP_011512460.1 14 - 15 2497 80
          XM_011514159.2 XP_011512461.1 11 - 12 5259 80
          XM_017010016.1 XP_016865505.1 14 - 15 2494 80
          GenBank mRNA AK293441.1 - 5 - 6 827 80
          BC115696.1 - 15 - 16 2372 40038868 80
          KJ534937.1 - 13 - 14 1928 80
          KJ535077.1 - 12 - 13 1720 80
          U52840.1 - 15 - 16 2565 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500010043.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05001151.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SEMA5A

          Panther Classification:

          Molecular Function -

          membrane-bound signaling molecule intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          neural crest cell migration
          positive regulation of endothelial cell proliferation
          blood vessel endothelial cell proliferation involved in sprouting angiogenesis
          cell adhesion
          negative regulation of cell adhesion
          cell-cell signaling
          nervous system development
          axonal fasciculation
          diencephalon development
          positive regulation of cell migration
          positive regulation of actin filament depolymerization
          positive regulation of catenin import into nucleus
          positive regulation of angiogenesis
          positive regulation of axon extension involved in axon guidance
          negative regulation of axon extension involved in axon guidance
          positive chemotaxis
          negative chemotaxis
          positive regulation of protein kinase B signaling
          cell chemotaxis
          semaphorin-plexin signaling pathway
          signal clustering
          negative regulation of endothelial cell apoptotic process
          positive regulation of endothelial cell chemotaxis
          semaphorin receptor binding
          chondroitin sulfate proteoglycan binding
          heparan sulfate proteoglycan binding
          chemorepellent activity
          syndecan binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05016491.hg.1 ENST00000382496.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016491.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05016491.hg.1 TC05001151.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500170613.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170613.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500170613.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170613.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500170613.hg.1 TC0500010043.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline