Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01549383_m1
          See other SEMA5A GE Assays ›
          Gene Symbol:
          SEMA5A
          Gene Name:
          semaphorin 5A
          Gene Aliases:
          SEMAF, semF
          Chromosome Location:
          Chr.5: 9035026 - 9546121 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01549383_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SEMA5A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003966.2 NP_003957.2 4 - 5 932 70
          XM_006714506.2 XP_006714569.1 5 - 6 1031 70
          XM_006714507.3 XP_006714570.1 3 - 4 594 70
          XM_011514155.2 XP_011512457.1 4 - 5 555 70
          XM_011514156.1 XP_011512458.1 6 - 7 1104 70
          XM_011514157.1 XP_011512459.1 5 - 6 959 70
          XM_011514158.1 XP_011512460.1 3 - 4 789 70
          GenBank mRNA BC115696.1 - 4 - 5 664 40038868 70
          KJ534937.1 - 2 - 3 220 70
          KJ535077.1 - 2 - 3 220 70
          U52840.1 - 4 - 5 857 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500010043.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05001151.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SEMA5A

          Panther Classification:

          Molecular Function -

          membrane-bound signaling molecule intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          neural crest cell migration
          positive regulation of endothelial cell proliferation
          blood vessel endothelial cell proliferation involved in sprouting angiogenesis
          cell adhesion
          negative regulation of cell adhesion
          cell-cell signaling
          nervous system development
          axonal fasciculation
          diencephalon development
          positive regulation of cell migration
          positive regulation of actin filament depolymerization
          positive regulation of catenin import into nucleus
          positive regulation of angiogenesis
          positive regulation of axon extension involved in axon guidance
          negative regulation of axon extension involved in axon guidance
          positive chemotaxis
          negative chemotaxis
          positive regulation of protein kinase B signaling
          cell chemotaxis
          semaphorin-plexin signaling pathway
          signal clustering
          negative regulation of endothelial cell apoptotic process
          positive regulation of endothelial cell chemotaxis
          semaphorin receptor binding
          chondroitin sulfate proteoglycan binding
          heparan sulfate proteoglycan binding
          chemorepellent activity
          syndecan binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05016506.hg.1 ENST00000382496.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016506.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05016506.hg.1 TC05001151.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 ENST00000514923.4 Ensembl
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay TC0500010043.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500170635.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170635.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170635.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170635.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500170635.hg.1 ENST00000382496.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170635.hg.1 ENST00000509486.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170635.hg.1 ENST00000513968.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170635.hg.1 NM_003966.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500170635.hg.1 TC0500010043.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline