Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256063.1 | NP_001242992.1 | 3 - 4 | 320 | 140 | |
NM_001256064.1 | NP_001242993.1 | 3 - 4 | 533 | 140 | ||
NM_001843.3 | NP_001834.2 | 3 - 4 | 320 | 140 | ||
XM_005268651.2 | XP_005268708.1 | 3 - 4 | 683 | 140 | ||
XM_006719241.2 | XP_006719304.1 | 3 - 4 | 307 | 140 | ||
XM_011537926.2 | XP_011536228.1 | 3 - 4 | 272 | 140 | ||
XM_011537927.2 | XP_011536229.1 | 3 - 4 | 329 | 140 | ||
XM_017018821.1 | XP_016874310.1 | 3 - 4 | 323 | 140 | ||
XM_017018822.1 | XP_016874311.1 | 3 - 4 | 272 | 140 | ||
XM_017018823.1 | XP_016874312.1 | 3 - 4 | 683 | 140 | ||
XM_017018824.1 | XP_016874313.1 | 3 - 4 | 307 | 140 | ||
XM_017018825.1 | XP_016874314.1 | 3 - 4 | 272 | 140 | ||
XM_017018826.1 | XP_016874315.1 | 3 - 4 | 307 | 140 | ||
XM_017018827.1 | XP_016874316.1 | 3 - 4 | 329 | 140 | ||
GenBank mRNA | AK289544.1 | - | 3 - 4 | 412 | 140 | |
AK289698.1 | - | 3 - 4 | 262 | 140 | ||
BC036569.2 | - | 3 - 4 | 259 | 5273941 | 140 | |
U07819.1 | - | 2 - 3 | 93 | 140 | ||
Z21488.1 | - | 3 - 4 | 214 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Probe spans exons |