Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01552925_m1
          See other FGFR2 GE Assays ›
          Gene Symbol:
          FGFR2
          Gene Name:
          fibroblast growth factor receptor 2
          Gene Aliases:
          BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
          Chromosome Location:
          Chr.10: 121478330 - 121598656 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01552925_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FGFR2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000141.4 NP_000132.3 4 - 5 1104 75
          NM_001144913.1 NP_001138385.1 3 - 4 607 75
          NM_001144914.1 NP_001138386.1 3 - 4 607 75
          NM_001144915.1 NP_001138387.1 3 - 4 509 75
          NM_001144917.1 NP_001138389.1 4 - 5 1104 75
          NM_001144919.1 NP_001138391.1 3 - 4 837 75
          NM_001320658.1 NP_001307587.1 4 - 5 1104 75
          NM_022970.3 NP_075259.4 4 - 5 1104 75
          NM_023029.2 NP_075418.1 2 - 3 340 75
          XM_006717708.2 XP_006717771.1 4 - 5 1302 75
          XM_006717710.3 XP_006717773.1 4 - 5 1302 75
          XM_006717711.2 XP_006717774.1 2 - 3 340 75
          XM_017015920.1 XP_016871409.1 4 - 5 1302 75
          XM_017015921.1 XP_016871410.1 4 - 5 1302 75
          XM_017015922.1 XP_016871411.1 2 - 3 340 75
          XM_017015923.1 XP_016871412.1 2 - 3 340 75
          GenBank mRNA AB030073.1 - 4 - 5 875 75
          AB030074.1 - 4 - 5 875 75
          AB030075.1 - 4 - 5 875 75
          AB030076.1 - 4 - 5 875 75
          AB030077.1 - 4 - 5 875 75
          AB030078.1 - 4 - 5 875 75
          AB084153.1 - 3 - 4 457 75
          AK026508.1 - 3 - 4 811 75
          AK294026.1 - 3 - 4 509 75
          EF057068.1 - 2 - 3 190 75
          EU826599.1 - 3 - 4 457 75
          EU826600.1 - 3 - 4 457 75
          KP192323.1 - 2 - 3 274 75
          KP192324.1 - 2 - 3 274 75
          KP192326.1 - 2 - 3 274 75
          KP192327.1 - 2 - 3 274 75
          KP192328.1 - 3 - 4 541 75
          KP192330.1 - 2 - 3 274 75
          KP192331.1 - 3 - 4 541 75
          M55614.1 - 3 - 4 469 75
          M80634.1 - 4 - 5 875 75
          M87770.1 - 4 - 5 730 75
          M87771.1 - 4 - 5 944 75
          M87772.1 - 4 - 5 732 75
          M97193.1 - 4 - 5 1068 75
          U11814.1 - 4 - 5 715 75
          X52832.1 - 4 - 5 636 75
          X56191.1 - 3 - 4 481 75
          Z71929.1 - 3 - 4 469 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000012050.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001711.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FGFR2

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          angiogenesis
          ureteric bud development
          in utero embryonic development
          epithelial to mesenchymal transition
          positive regulation of mesenchymal cell proliferation
          outflow tract septum morphogenesis
          membranous septum morphogenesis
          apoptotic process
          cell-cell signaling
          axonogenesis
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          regulation of smoothened signaling pathway
          post-embryonic development
          embryonic pattern specification
          organ morphogenesis
          regulation of cell fate commitment
          positive regulation of phospholipase activity
          regulation of phosphatidylinositol 3-kinase signaling
          morphogenesis of embryonic epithelium
          peptidyl-tyrosine phosphorylation
          orbitofrontal cortex development
          ventricular zone neuroblast division
          pyramidal neuron development
          gland morphogenesis
          positive regulation of Wnt signaling pathway
          bone mineralization
          lung development
          epithelial cell differentiation
          midbrain development
          otic vesicle formation
          hair follicle morphogenesis
          lacrimal gland development
          regulation of osteoblast proliferation
          multicellular organism growth
          organ growth
          fibroblast growth factor receptor signaling pathway involved in negative regulation of apoptotic process in bone marrow
          fibroblast growth factor receptor signaling pathway involved in hemopoiesis
          fibroblast growth factor receptor signaling pathway involved in positive regulation of cell proliferation in bone marrow
          fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
          phosphatidylinositol-3-phosphate biosynthetic process
          regulation of multicellular organism growth
          regulation of fibroblast growth factor receptor signaling pathway
          inner ear morphogenesis
          odontogenesis
          positive regulation of MAPK cascade
          positive regulation of GTPase activity
          cell fate commitment
          regulation of osteoblast differentiation
          positive regulation of cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          lung alveolus development
          mesodermal cell differentiation
          embryonic digestive tract morphogenesis
          embryonic organ morphogenesis
          digestive tract development
          embryonic organ development
          reproductive structure development
          positive regulation of smooth muscle cell proliferation
          embryonic cranial skeleton morphogenesis
          skeletal system morphogenesis
          epidermis morphogenesis
          branching morphogenesis of a nerve
          mesenchymal cell differentiation
          positive regulation of epithelial cell proliferation
          negative regulation of epithelial cell proliferation
          regulation of smooth muscle cell differentiation
          positive regulation of cell division
          ventricular cardiac muscle tissue morphogenesis
          positive regulation of cardiac muscle cell proliferation
          limb bud formation
          bone development
          bone morphogenesis
          branching involved in prostate gland morphogenesis
          branching involved in salivary gland morphogenesis
          bud elongation involved in lung branching
          lung lobe morphogenesis
          lung-associated mesenchyme development
          positive regulation of epithelial cell proliferation involved in lung morphogenesis
          prostate gland morphogenesis
          prostate epithelial cord elongation
          prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
          squamous basal epithelial stem cell differentiation involved in prostate gland acinus development
          fibroblast growth factor receptor signaling pathway involved in mammary gland specification
          lateral sprouting from an epithelium
          mammary gland bud formation
          epithelial cell proliferation involved in salivary gland morphogenesis
          branch elongation involved in salivary gland morphogenesis
          branching involved in labyrinthine layer morphogenesis
          regulation of branching involved in prostate gland morphogenesis
          regulation of morphogenesis of a branching structure
          mesenchymal cell differentiation involved in lung development
          mesenchymal cell proliferation involved in lung development
          regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of canonical Wnt signaling pathway
          protein tyrosine kinase activity
          fibroblast growth factor-activated receptor activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          ATP binding
          heparin binding
          1-phosphatidylinositol-3-kinase activity
          fibroblast growth factor binding
          protein homodimerization activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 ENST00000346997.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 ENST00000351936.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 ENST00000356226.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 ENST00000357555.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 ENST00000358487.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 ENST00000359354.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 ENST00000360144.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 ENST00000369056.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 ENST00000369059.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 ENST00000369060.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 ENST00000369061.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 ENST00000369062.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 ENST00000457416.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 NM_000141.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 NM_001144913.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 NM_001144914.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 NM_001144915.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 NM_001144916.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 NM_001144917.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 NM_001144918.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 NM_001144919.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 NM_022970.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10022622.hg.1 TC10001711.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000336553.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000346997.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000351936.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000356226.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000357555.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000358487.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000359354.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000360144.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000369056.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000369058.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000369059.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000369060.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000369061.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000457416.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000490349.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000604236.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000613048.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_000141.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144913.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144915.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144916.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144917.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144918.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144919.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_022970.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_023029.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NR_073009.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000336553.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000346997.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000351936.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000356226.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000357555.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000358487.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000359354.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000360144.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000369056.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000369058.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000369059.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000369060.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000369061.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000457416.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000490349.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000604236.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 ENST00000613048.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_000141.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144913.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144915.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144916.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144917.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144918.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_001144919.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_022970.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NM_023029.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 NR_073009.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182429.hg.1 TC1000012050.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline