Search
Search

ARHGEF10L| Gene Symbol: ARHGEF10L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011722.2 | NP_001011722.2 | 25 - 26 | 3110 | 56 | |
| NM_001319837.1 | NP_001306766.1 | 24 - 25 | 3095 | 56 | ||
| NM_001319838.1 | NP_001306767.1 | 16 - 17 | 2334 | 56 | ||
| NM_001328124.1 | NP_001315053.1 | 19 - 20 | 2559 | 56 | ||
| NM_018125.3 | NP_060595.3 | 27 - 28 | 3343 | 56 | ||
| NR_137287.1 | 28 - 29 | 4090 | 56 | |||
| NR_137288.1 | 27 - 28 | 3970 | 56 | |||
| XM_005245923.2 | XP_005245980.2 | 26 - 27 | 3202 | 56 | ||
| XM_005245925.2 | XP_005245982.2 | 26 - 27 | 3190 | 56 | ||
| XM_005245927.2 | XP_005245984.2 | 19 - 20 | 2559 | 56 | ||
| XM_005245929.2 | XP_005245986.2 | 18 - 19 | 2544 | 56 | ||
| XM_006710728.1 | XP_006710791.1 | 26 - 27 | 3384 | 56 | ||
| XM_006710729.1 | XP_006710792.1 | 26 - 27 | 3199 | 56 | ||
| XM_006710731.1 | XP_006710794.1 | 26 - 27 | 3381 | 56 | ||
| XM_011541691.2 | XP_011539993.1 | 25 - 26 | 3199 | 56 | ||
| XM_011541692.2 | XP_011539994.1 | 24 - 25 | 3193 | 56 | ||
| XM_011541693.2 | XP_011539995.1 | 24 - 25 | 3184 | 56 | ||
| XM_017001617.1 | XP_016857106.1 | 25 - 26 | 3175 | 56 | ||
| XM_017001618.1 | XP_016857107.1 | 20 - 21 | 2477 | 56 | ||
| XM_017001621.1 | XP_016857110.1 | 26 - 27 | 3326 | 56 | ||
| XM_017001622.1 | XP_016857111.1 | 25 - 26 | 3311 | 56 | ||
| GenBank mRNA | AB046846.1 | - | 27 - 28 | 3343 | 56 | |
| AK092020.1 | - | 16 - 17 | 2331 | 56 | ||
| AK126877.1 | - | 18 - 19 | 3449 | 56 | ||
| AK300264.1 | - | 18 - 19 | 2507 | 56 | ||
| BC065561.1 | - | 13 - 14 | 1845 | 56 | ||
| BC080596.1 | - | 16 - 17 | 2006 | 56 | ||
| BC117171.1 | - | 25 - 26 | 3100 | 40125722 | 56 | |
| BC143347.1 | - | 25 - 26 | 3100 | 9051852 | 56 | |
| DQ139833.1 | - | 24 - 25 | 3052 | 56 | ||
Set Membership: |
|