Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01554038_m1
          See other CCNY GE Assays ›
          Gene Symbol:
          CCNY
          Gene Name:
          cyclin Y
          Gene Aliases:
          C10orf9, CBCP1, CCNX, CFP1
          Chromosome Location:
          Chr.10: 35246981 - 35572670 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01554038_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCNY
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001282852.1 NP_001269781.1 4 - 5 506 138
          NM_001282853.1 NP_001269782.1 6 - 7 644 138
          NM_145012.5 NP_659449.3 5 - 6 581 138
          NM_181698.3 NP_859049.2 7 - 8 605 138
          XM_006717392.3 XP_006717455.1 5 - 6 350 138
          XM_011519358.1 XP_011517660.1 8 - 9 672 138
          XM_011519360.2 XP_011517662.1 9 - 10 774 138
          XM_011519361.1 XP_011517663.1 9 - 10 656 138
          XM_011519362.1 XP_011517664.1 8 - 9 636 138
          XM_011519363.1 XP_011517665.1 8 - 9 649 138
          XM_011519364.1 XP_011517666.1 10 - 11 770 138
          XM_011519365.2 XP_011517667.1 9 - 10 689 138
          XM_011519366.1 XP_011517668.1 8 - 9 619 138
          XM_011519367.1 XP_011517669.1 6 - 7 449 138
          XM_011519368.2 XP_011517670.1 5 - 6 344 138
          XM_017015839.1 XP_016871328.1 5 - 6 267 138
          XM_017015840.1 XP_016871329.1 5 - 6 269 138
          XM_017015841.1 XP_016871330.1 5 - 6 267 138
          GenBank mRNA AF429969.1 - 7 - 8 561 138
          AF465728.1 - 5 - 6 327 138
          AK057280.1 - 7 - 8 561 138
          AK314666.1 - 6 - 7 620 138
          AL834355.1 - 5 - 6 439 138
          BC069224.1 - 5 - 6 407 138
          BC094815.1 - 5 - 6 433 6340750 138
          BC104773.1 - 6 - 7 390 8143776 138
          BC104801.1 - 6 - 7 390 8143804 138
          BC143450.1 - 5 - 6 283 9051958 138
          BC143455.1 - 6 - 7 390 9051963 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC10000240.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCNY

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          Wnt signaling pathway
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          cell division
          regulation of canonical Wnt signaling pathway
          protein binding
          cyclin-dependent protein serine/threonine kinase regulator activity
          protein kinase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10002793.hg.1 ENST00000265375.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10002793.hg.1 ENST00000339497.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10002793.hg.1 ENST00000374704.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10002793.hg.1 ENST00000374706.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10002793.hg.1 ENST00000456784.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10002793.hg.1 ENST00000492478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10002793.hg.1 ENST00000537547.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10002793.hg.1 NM_145012.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10002793.hg.1 NM_181698.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10002793.hg.1 TC10000240.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline