Search
Search

EPB41L5| Gene Symbol: EPB41L5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330310.2 | NP_001317239.1 | 23 - 24 | 2143 | 72 | |
| GenBank mRNA | AB046768.1 | - | 16 - 17 | 1441 | 72 | |
Set Membership: |
|