Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01555466_m1
          See other ATG5 GE Assays ›
          Gene Symbol:
          ATG5
          Gene Name:
          autophagy related 5
          Gene Aliases:
          APG5, APG5-LIKE, APG5L, ASP, hAPG5
          Chromosome Location:
          Chr.6: 106184476 - 106325820 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01555466_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ATG5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286107.1 NP_001273036.1 2 - 3 458 92
          NR_104403.1 1 - 2 163 92
          GenBank mRNA AF293841.1 - 1 - 2 105 92
          AK299577.1 - 2 - 3 450 92
          CR749386.1 - 2 - 3 377 92
          JQ918354.1 - 1 - 2 105 92

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF293841.1, AK299577.1, CR749386.1, NM_001286107.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 Apoptosis Homo sapiens Probe spans exons

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          mitophagy
          blood vessel remodeling
          regulation of cytokine secretion involved in immune response
          C-terminal protein lipidation
          autophagy
          apoptotic process
          cellular response to nitrogen starvation
          response to fungus
          macroautophagy
          cellular homeostasis
          antigen processing and presentation of endogenous antigen
          negative regulation of protein ubiquitination
          aggrephagy
          negative stranded viral RNA replication
          vasodilation
          response to drug
          negative regulation of apoptotic process
          post-translational protein modification
          nucleophagy
          negative thymic T cell selection
          innate immune response
          otolith development
          regulation of release of sequestered calcium ion into cytosol
          ventricular cardiac muscle cell development
          heart contraction
          negative regulation of cell death
          protein lipidation involved in autophagosome assembly
          positive regulation of mucus secretion
          regulation of cilium assembly
          positive regulation of mitophagy
          negative regulation of reactive oxygen species metabolic process
          negative regulation of histone H4-K16 acetylation
          protein binding
          Atg8 ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000343245.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000360666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369070.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000369076.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000475645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000476518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 ENST00000613993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286106.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286108.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_001286111.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NM_004849.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104402.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012731.hg.1 NR_104403.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline