| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000517327.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000517327.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000517327.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000517327.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000519777.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000519777.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000519777.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
ENST00000519777.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018033.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018033.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08018033.hg.1 |
TC08001392.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800174949.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800174949.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800174949.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800174949.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800174949.hg.1 |
TC0800011023.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000320005.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000517327.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000517327.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000517327.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000517327.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000519777.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000519777.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000519777.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
ENST00000519777.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011023.hg.1 |
NM_019098.4 |
Refseq |