Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01559335_g1
          See other TRIM3 GE Assays ›
          Gene Symbol:
          TRIM3
          Gene Name:
          tripartite motif containing 3
          Gene Aliases:
          BERP, HAC1, RNF22, RNF97
          Chromosome Location:
          Chr.11: 6448613 - 6474459 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01559335_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRIM3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001248006.2 NP_001234935.1 4 - 5 909 159
          NM_001248007.2 NP_001234936.1 3 - 4 529 159
          NM_006458.4 NP_006449.2 5 - 6 867 159
          NM_033278.4 NP_150594.2 4 - 5 702 159
          XM_017017098.3 XP_016872587.1 4 - 5 728 159
          XM_024448328.2 XP_024304096.1 5 - 6 769 159
          XM_047426257.1 XP_047282213.1 5 - 6 853 159
          XM_047426258.1 XP_047282214.1 4 - 5 705 159
          XM_047426259.1 XP_047282215.1 4 - 5 569 159
          XM_047426261.1 XP_047282217.1 3 - 4 534 159
          XM_047426262.1 XP_047282218.1 4 - 5 695 159
          GenBank mRNA AF045239.1 - 3 - 4 514 159
          AF220020.1 - 3 - 4 514 159
          AF220021.1 - 4 - 5 481 126
          AF220022.1 - 3 - 4 514 159
          AK291396.1 - 4 - 5 909 159
          AK298843.1 - 3 - 4 448 159
          AK299586.1 - 4 - 5 702 159
          AK311127.1 - 4 - 5 700 159
          AL360192.1 - 4 - 5 451 126
          BC096827.1 - 4 - 5 745 159
          BM758857.1 - 2 - 3 197 159

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TRIM3

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          toll-like receptor signaling pathway
          nervous system development
          protein transport
          protein ubiquitination
          positive regulation of toll-like receptor 3 signaling pathway
          proteasome-mediated ubiquitin-dependent protein catabolic process
          neural precursor cell proliferation
          protein K63-linked ubiquitination
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          transferase activity
          identical protein binding
          metal ion binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline