| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014103.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198137.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198137.hg.1 |
ENST00000375832.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198137.hg.1 |
TC0600014103.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600198139.hg.1 |
TC0600014103.hg.1 |
Clariom D TC |