Search
Search

ABCC1| Gene Symbol: ABCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004996.3 | NP_004987.2 | 26 - 27 | 3994 | 68 | |
| XM_011522497.1 | XP_011520799.1 | 26 - 27 | 3810 | 68 | ||
| XM_011522498.2 | XP_011520800.1 | 25 - 26 | 3729 | 68 | ||
| XM_017023237.1 | XP_016878726.1 | 26 - 27 | 3876 | 68 | ||
| XM_017023238.1 | XP_016878727.1 | 25 - 26 | 3750 | 68 | ||
| XM_017023239.1 | XP_016878728.1 | 25 - 26 | 3738 | 68 | ||
| XM_017023240.1 | XP_016878729.1 | 25 - 26 | 3699 | 68 | ||
| XM_017023241.1 | XP_016878730.1 | 24 - 25 | 3612 | 68 | ||
| XM_017023242.1 | XP_016878731.1 | 24 - 25 | 3531 | 68 | ||
| GenBank mRNA | AB209120.1 | - | 25 - 26 | 3544 | 68 | |
| BC001636.1 | - | 2 - 3 | 175 | 68 | ||
| BX648866.1 | - | 4 - 5 | 484 | 68 | ||
| CR749835.1 | - | 23 - 24 | 3302 | 68 | ||
| HQ917067.1 | - | 3 - 4 | 357 | 68 | ||
| L05628.1 | - | 26 - 27 | 4015 | 68 | ||
Set Membership: |
|