Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01567090_m1
          See other FGFR2 GE Assays ›
          Gene Symbol:
          FGFR2
          Gene Name:
          fibroblast growth factor receptor 2
          Gene Aliases:
          BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
          Chromosome Location:
          Chr.10: 121478330 - 121598656 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01567090_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FGFR2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001144913.1 NP_001138385.1 6 - 7 1088 63
          NM_001144919.1 NP_001138391.1 6 - 7 1318 63
          NM_022970.3 NP_075259.4 7 - 8 1585 63
          NR_073009.1 5 - 6 1240 63
          XM_006717708.2 XP_006717771.1 7 - 8 1783 63
          XM_006717710.3 XP_006717773.1 7 - 8 1783 63
          XM_006717711.2 XP_006717774.1 5 - 6 821 63
          XM_006717712.2 XP_006717775.1 4 - 5 743 63
          XM_017015920.1 XP_016871409.1 7 - 8 1783 63
          XM_017015922.1 XP_016871411.1 5 - 6 821 63
          GenBank mRNA AB030073.1 - 7 - 8 1356 63
          AB030074.1 - 7 - 8 1356 63
          AB030075.1 - 7 - 8 1356 63
          AB030076.1 - 7 - 8 1356 63
          AB030077.1 - 7 - 8 1356 63
          AB030078.1 - 7 - 8 1356 63
          AK026508.1 - 6 - 7 1292 63
          KP192323.1 - 5 - 6 755 63
          KP192324.1 - 5 - 6 755 63
          M80634.1 - 7 - 8 1356 63
          M97193.1 - 7 - 8 1549 63
          U11814.1 - 7 - 8 1196 63
          X56191.1 - 6 - 7 962 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000012050.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001711.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FGFR2

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          angiogenesis
          ureteric bud development
          in utero embryonic development
          epithelial to mesenchymal transition
          positive regulation of mesenchymal cell proliferation
          outflow tract septum morphogenesis
          membranous septum morphogenesis
          apoptotic process
          cell-cell signaling
          axonogenesis
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          regulation of smoothened signaling pathway
          post-embryonic development
          embryonic pattern specification
          organ morphogenesis
          regulation of cell fate commitment
          positive regulation of phospholipase activity
          regulation of phosphatidylinositol 3-kinase signaling
          morphogenesis of embryonic epithelium
          peptidyl-tyrosine phosphorylation
          orbitofrontal cortex development
          ventricular zone neuroblast division
          pyramidal neuron development
          gland morphogenesis
          positive regulation of Wnt signaling pathway
          bone mineralization
          lung development
          epithelial cell differentiation
          midbrain development
          otic vesicle formation
          hair follicle morphogenesis
          lacrimal gland development
          regulation of osteoblast proliferation
          multicellular organism growth
          organ growth
          fibroblast growth factor receptor signaling pathway involved in negative regulation of apoptotic process in bone marrow
          fibroblast growth factor receptor signaling pathway involved in hemopoiesis
          fibroblast growth factor receptor signaling pathway involved in positive regulation of cell proliferation in bone marrow
          fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
          phosphatidylinositol-3-phosphate biosynthetic process
          regulation of multicellular organism growth
          regulation of fibroblast growth factor receptor signaling pathway
          inner ear morphogenesis
          odontogenesis
          positive regulation of MAPK cascade
          positive regulation of GTPase activity
          cell fate commitment
          regulation of osteoblast differentiation
          positive regulation of cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          lung alveolus development
          mesodermal cell differentiation
          embryonic digestive tract morphogenesis
          embryonic organ morphogenesis
          digestive tract development
          embryonic organ development
          reproductive structure development
          positive regulation of smooth muscle cell proliferation
          embryonic cranial skeleton morphogenesis
          skeletal system morphogenesis
          epidermis morphogenesis
          branching morphogenesis of a nerve
          mesenchymal cell differentiation
          positive regulation of epithelial cell proliferation
          negative regulation of epithelial cell proliferation
          regulation of smooth muscle cell differentiation
          positive regulation of cell division
          ventricular cardiac muscle tissue morphogenesis
          positive regulation of cardiac muscle cell proliferation
          limb bud formation
          bone development
          bone morphogenesis
          branching involved in prostate gland morphogenesis
          branching involved in salivary gland morphogenesis
          bud elongation involved in lung branching
          lung lobe morphogenesis
          lung-associated mesenchyme development
          positive regulation of epithelial cell proliferation involved in lung morphogenesis
          prostate gland morphogenesis
          prostate epithelial cord elongation
          prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
          squamous basal epithelial stem cell differentiation involved in prostate gland acinus development
          fibroblast growth factor receptor signaling pathway involved in mammary gland specification
          lateral sprouting from an epithelium
          mammary gland bud formation
          epithelial cell proliferation involved in salivary gland morphogenesis
          branch elongation involved in salivary gland morphogenesis
          branching involved in labyrinthine layer morphogenesis
          regulation of branching involved in prostate gland morphogenesis
          regulation of morphogenesis of a branching structure
          mesenchymal cell differentiation involved in lung development
          mesenchymal cell proliferation involved in lung development
          regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of canonical Wnt signaling pathway
          protein tyrosine kinase activity
          fibroblast growth factor-activated receptor activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          ATP binding
          heparin binding
          1-phosphatidylinositol-3-kinase activity
          fibroblast growth factor binding
          protein homodimerization activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10022611.hg.1 ENST00000360144.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022611.hg.1 ENST00000369056.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022611.hg.1 ENST00000369059.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022611.hg.1 ENST00000457416.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10022611.hg.1 NM_001144913.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10022611.hg.1 NM_001144919.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10022611.hg.1 NM_022970.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10022611.hg.1 TC10001711.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000360144.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000369056.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000369058.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000369059.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000457416.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000463870.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000490349.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000604236.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 NM_001144913.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 NM_001144919.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 NM_022970.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 NR_073009.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000360144.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000369056.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000369058.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000369059.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000457416.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000463870.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000490349.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 ENST00000604236.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 NM_001144913.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 NM_001144919.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 NM_022970.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 NR_073009.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000182419.hg.1 TC1000012050.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline