| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05018765.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05018765.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05018765.hg.1 |
TC05001329.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174510.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500174510.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174510.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500174510.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500174510.hg.1 |
TC0500010662.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |