Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01572638_m1
          See other SLC8A3 GE Assays ›
          Gene Symbol:
          SLC8A3
          Gene Name:
          solute carrier family 8 member A3
          Gene Aliases:
          NCX3
          Chromosome Location:
          Chr.14: 70044217 - 70189626 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01572638_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC8A3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033262.4 NP_150287.1 4 - 5 2657 76
          GenBank mRNA AJ304852.1 - 3 - 4 1965 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400009568.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC8A3

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          ion transport
          mitochondrial calcium ion transport
          cellular calcium ion homeostasis
          cell communication
          learning
          memory
          regulation of skeletal muscle contraction
          telencephalon development
          sodium ion transmembrane transport
          myelination
          oligodendrocyte differentiation
          mitochondrial calcium ion homeostasis
          long-term synaptic potentiation
          calcium ion transport into cytosol
          calcium ion transmembrane transport
          cellular response to cAMP
          cellular response to hypoxia
          regulation of cardiac conduction
          calcium ion export from cell
          calcium ion import into cell
          calcium:sodium antiporter activity
          calmodulin binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000216568.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000356921.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000357887.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000381269.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000394330.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000494208.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000528359.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000533541.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000534137.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NM_001130417.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NM_033262.4 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NM_058240.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NM_182932.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NM_182936.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NM_183002.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NR_104122.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000216568.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000356921.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000357887.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000381269.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000394330.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000494208.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000528359.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000533541.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 ENST00000534137.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NM_001130417.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NM_033262.4 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NM_058240.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NM_182932.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NM_182936.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NM_183002.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 NR_104122.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400170523.hg.1 TC1400009568.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline