Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01573118_m1
          See other ARFGEF2 GE Assays ›
          Gene Symbol:
          ARFGEF2
          Gene Name:
          ADP ribosylation factor guanine nucleotide exchange factor 2
          Gene Aliases:
          BIG2, PVNH2, dJ1164I10.1
          Chromosome Location:
          Chr.20: 48921721 - 49036693 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01573118_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ARFGEF2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006420.2 NP_006411.2 33 - 34 4659 110
          XM_005260252.3 XP_005260309.1 33 - 34 4673 110
          GenBank mRNA AB209397.1 - 13 - 14 1615 110
          AF084521.1 - 33 - 34 4507 110
          AK022623.1 - 12 - 13 1481 110
          AK074303.1 - 16 - 17 2035 110
          AK302526.1 - 19 - 20 2514 110
          BC144625.1 - 32 - 33 4689 110
          BC144626.1 - 34 - 35 5444 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000007620.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000382.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ARFGEF2

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          receptor recycling
          exocytosis
          Golgi to plasma membrane transport
          endosome organization
          endomembrane system organization
          protein transport
          regulation of ARF protein signal transduction
          positive regulation of tumor necrosis factor production
          intracellular signal transduction
          positive regulation of GTPase activity
          guanyl-nucleotide exchange factor activity
          ARF guanyl-nucleotide exchange factor activity
          protein binding
          myosin binding
          protein kinase A regulatory subunit binding
          GABA receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000493140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000493140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 ENST00000493140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20005524.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20005524.hg.1 NM_006420.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20005524.hg.1 TC20000382.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000493140.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000493140.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 ENST00000493140.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay TC2000007620.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000153132.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000153132.hg.1 NM_006420.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000153132.hg.1 TC2000007620.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000153132.hg.1 ENST00000371917.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000153132.hg.1 NM_006420.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline