Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01584228_g1
          See other ARHGEF19 GE Assays ›
          Gene Symbol:
          ARHGEF19
          Gene Name:
          Rho guanine nucleotide exchange factor 19
          Gene Aliases:
          WGEF
          Chromosome Location:
          Chr.1: 16197856 - 16212699 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01584228_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ARHGEF19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153213.3 NP_694945.2 5 - 6 1009 111
          XM_011540706.2 XP_011539008.1 6 - 7 1173 111
          GenBank mRNA AK091281.1 - 5 - 6 1045 111
          AK297707.1 - 5 - 6 1071 111
          BC012982.2 - 4 - 5 585 111
          BC040640.1 - 5 - 6 1009 3847806 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100013034.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002259.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ARHGEF19

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of actin cytoskeleton organization
          regulation of Rho protein signal transduction
          wound healing
          positive regulation of GTPase activity
          Wnt signaling pathway, planar cell polarity pathway
          Rho guanyl-nucleotide exchange factor activity
          GTPase activator activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01034483.hg.1 ENST00000270747.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01034483.hg.1 ENST00000375607.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01034483.hg.1 ENST00000421561.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01034483.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01034483.hg.1 TC01002259.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000441785.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000441785.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000441785.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000441785.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000471928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000471928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000471928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000471928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000471928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000471928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478210.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478210.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478210.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478210.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 ENST00000478210.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000441785.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000441785.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000441785.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000441785.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000449495.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000471928.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000471928.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000471928.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000471928.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000471928.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000471928.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478117.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478210.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478210.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478210.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478210.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 ENST00000478210.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013034.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100196061.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196061.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100196061.hg.1 ENST00000270747.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196061.hg.1 NM_153213.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100196061.hg.1 TC0100013034.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline