Search
Search

MAP2K7| Gene Symbol: MAP2K7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297555.1 | NP_001284484.1 | 2 - 3 | 339 | 113 | |
| XM_006722800.2 | XP_006722863.1 | 2 - 3 | 334 | 113 | ||
| GenBank mRNA | AK300988.1 | - | 2 - 3 | 234 | 113 | |
| DQ445915.1 | - | 2 - 3 | 174 | 113 | ||
Set Membership: |
|