Search
Search
| Gene Symbol: NRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261463.1 | NP_001248392.1 | 33 - 34 | 3997 | 138 | |
| NM_001322945.1 | NP_001309874.1 | 32 - 33 | 3889 | 138 | ||
| NM_006175.4 | NP_006166.3 | 32 - 33 | 3892 | 138 | ||
| NM_198060.3 | NP_932326.2 | 33 - 34 | 3997 | 138 | ||
| XM_005269864.2 | XP_005269921.1 | 32 - 33 | 3868 | 138 | ||
| XM_005269865.2 | XP_005269922.1 | 32 - 33 | 3865 | 138 | ||
| XM_005269867.2 | XP_005269924.1 | 33 - 34 | 3973 | 138 | ||
| XM_006717870.2 | XP_006717933.1 | 32 - 33 | 3868 | 138 | ||
| XM_011539832.2 | XP_011538134.1 | 33 - 34 | 3973 | 138 | ||
| GenBank mRNA | AL831980.2 | - | 33 - 34 | 3924 | 138 | |
| AL832025.2 | - | 33 - 34 | 3924 | 138 | ||
| AL832330.2 | - | 33 - 34 | 3924 | 138 | ||
| AL832457.3 | - | 19 - 20 | 2350 | 138 | ||
| AY081943.2 | - | 32 - 33 | 3869 | 138 | ||
| AY177620.1 | - | 33 - 34 | 3950 | 138 | ||
| AY177621.1 | - | 32 - 33 | 3845 | 138 | ||
| BC126407.1 | - | 33 - 34 | 3833 | 8992123 | 138 | |
| BC143751.1 | - | 33 - 34 | 3833 | 9052265 | 138 | |
| BX640682.2 | - | 33 - 34 | 3924 | 138 | ||
| BX640730.1 | - | 33 - 34 | 4081 | 138 | ||
| BX641052.1 | - | 33 - 34 | 3961 | 138 | ||
| BX647658.1 | - | 33 - 34 | 3998 | 138 | ||
Set Membership: |
|