| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022073.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022073.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022073.hg.1 |
TC02001398.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022072.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022072.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022072.hg.1 |
TC02001398.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000474529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000474529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000474529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000474529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000474529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000474529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000485563.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000485563.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
ENST00000485563.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000474529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000474529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000474529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000474529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000474529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000474529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000485563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000485563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
ENST00000485563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011093.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179709.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179709.hg.1 |
ENST00000485563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179709.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179709.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179709.hg.1 |
ENST00000485563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179709.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179709.hg.1 |
TC0200011093.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200179707.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179707.hg.1 |
ENST00000485563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179707.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179707.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179707.hg.1 |
ENST00000485563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179707.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179707.hg.1 |
TC0200011093.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200179708.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179708.hg.1 |
ENST00000485563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179708.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179708.hg.1 |
ENST00000392027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179708.hg.1 |
ENST00000485563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200179708.hg.1 |
NM_001632.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200179708.hg.1 |
TC0200011093.hg.1 |
Clariom D TC |