Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01661623_s1
          See other KRTAP2-4 GE Assays ›
          Gene Symbol:
          KRTAP2-4 KRTAP2-2
          Gene Name:
          keratin associated protein 2-4
          keratin associated protein 2-2
          Gene Aliases:
          KAP2.1B, KAP2.4, KRTAP2.4
          KAP2.2
          Chromosome Location:
          Chr.17: 41065116 - 41065879 on Build GRCh38
          Chromosome Location:
          Chr.17: 41054498 - 41055211 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01661623_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KRTAP2-4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AJ406929.1 - 358 86
          Gene Symbol: KRTAP2-2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033032.2 NP_149021.2 1 537 86
          GenBank mRNA AJ302536.1 - 424 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700010652.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001484.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3286196 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2944 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_2943 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 9212 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P153022 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3286196 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3286196 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P131189 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8019575 - -
          Affymetrix Human Gene 1.0 ST Array 8015209 - -
          Affymetrix Human Gene 1.0 ST Array 8019577 - -
          Affymetrix Human Gene 1.0 ST Array 8015212 - -
          Affymetrix Human Gene 1.0 ST Array 8015216 - -
          Affymetrix Human Gene 1.0 ST Array 8015207 - -
          Affymetrix Human Gene 1.1 ST Array 8019576 - -
          Affymetrix Human Gene 1.1 ST Array 8019574 - -
          Affymetrix Human Gene 1.1 ST Array 8015206 - -
          Affymetrix Human Gene 1.1 ST Array 8015208 - -
          Affymetrix Human Gene 1.1 ST Array 8015210 - -
          Affymetrix Human Gene 1.1 ST Array 8015214 - -
          Affymetrix Human Transcriptome Array 2.0 TC1700010652.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010652.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010652.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010652.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010652.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010652.hg.1 ENST00000542910.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010652.hg.1 ENST00000542910.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010652.hg.1 NM_033032.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010652.hg.1 NM_033032.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010652.hg.1 NM_033032.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010652.hg.1 NM_033032.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010652.hg.1 NM_033032.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019695.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019695.hg.1 NM_033032.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019695.hg.1 TC17001484.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17019693.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019693.hg.1 NM_033032.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019693.hg.1 TC17001484.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17019694.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019694.hg.1 ENST00000542910.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019694.hg.1 NM_033032.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019694.hg.1 TC17001484.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17019696.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019696.hg.1 ENST00000542910.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019696.hg.1 NM_033032.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019696.hg.1 TC17001484.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700184194.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184194.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700184194.hg.1 TC1700010652.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184194.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184194.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700184195.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184195.hg.1 ENST00000542910.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184195.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700184195.hg.1 TC1700010652.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184195.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184195.hg.1 ENST00000542910.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184195.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700184198.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184198.hg.1 ENST00000542910.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184198.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700184198.hg.1 TC1700010652.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184198.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184198.hg.1 ENST00000542910.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184198.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay TC1700010652.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010652.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010652.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010652.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010652.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010652.hg.1 ENST00000542910.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010652.hg.1 ENST00000542910.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010652.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay TC1700010652.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay TC1700010652.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay TC1700010652.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay TC1700010652.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700184196.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184196.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700184196.hg.1 TC1700010652.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184196.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184196.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700184197.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184197.hg.1 NM_033032.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700184197.hg.1 TC1700010652.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184197.hg.1 ENST00000398477.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184197.hg.1 NM_033032.2 Refseq
          Human Genome U133 Plus 2.0 Array 234772_s_at - -
          Human Genome U133 Plus 2.0 Array 234772_PM_s_at - -
          Human Genome U133 Set 234772_s_at - -
          Human Genome U133 Set 234772_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008311 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008311 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1676146 NM_033032.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1676146 AJ302536.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1780344 NM_033032.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1780344 AJ302536.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline