| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009694.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009694.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009694.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009694.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009694.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009694.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009694.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009694.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009694.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009694.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009694.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009694.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012678.hg.1 |
ENST00000421715.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012678.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012678.hg.1 |
TC09000950.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012216.hg.1 |
ENST00000413767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012216.hg.1 |
ENST00000413767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012216.hg.1 |
ENST00000413767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012216.hg.1 |
ENST00000413767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012216.hg.1 |
NM_021268.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012216.hg.1 |
NM_021268.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012216.hg.1 |
NM_021268.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012216.hg.1 |
NM_021268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009694.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009694.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009694.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009694.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009694.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009694.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009694.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009694.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009694.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009694.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009694.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009694.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012216.hg.1 |
ENST00000413767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012216.hg.1 |
ENST00000413767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012216.hg.1 |
ENST00000413767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012216.hg.1 |
ENST00000413767.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012216.hg.1 |
NM_021268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012216.hg.1 |
NM_021268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012216.hg.1 |
NM_021268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012216.hg.1 |
NM_021268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166322.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166322.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166322.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166322.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166322.hg.1 |
TC0900009694.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900166321.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166321.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166321.hg.1 |
ENST00000421715.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166321.hg.1 |
NM_021068.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166321.hg.1 |
TC0900009694.hg.1 |
Clariom D TC |