Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01697229_g1
          See other ACTR3BP2 GE Assays ›
          Gene Symbol:
          ACTR3BP2
          Gene Name:
          ACTR3B pseudogene 2
          Gene Aliases:
          FKSG73
          Chromosome Location:
          Chr.2: 91941133 - 91942470 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01697229_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ACTR3BP2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027714.1 1 1218 125
          GenBank mRNA AY026351.1 - 1 1218 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200008431.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000565.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 198534 AK027641.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 29221 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3816042 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 34354 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 60859 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P12433787 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3816042 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3816042 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P141437 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P141437 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7927098 - -
          Affymetrix Human Gene 1.0 ST Array 8043488 - -
          Affymetrix Human Gene 1.0 ST Array 8001071 - -
          Affymetrix Human Gene 1.0 ST Array 8071060 - -
          Affymetrix Human Gene 1.0 ST Array 8071059 - -
          Affymetrix Human Gene 1.1 ST Array 8071058 - -
          Affymetrix Human Gene 1.1 ST Array 7927097 - -
          Affymetrix Human Gene 1.1 ST Array 8043487 - -
          Affymetrix Human Gene 1.1 ST Array 8001070 - -
          Affymetrix HT HG-U133+ PM Array Plate 224426_PM_s_at - -
          Affymetrix HT HG-U133+ PM Array Plate 224425_PM_x_at - -
          Affymetrix HT HG-U133+ PM Array Plate 224424_PM_x_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2200006476.hg.1 ENST00000623630.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC02000565.hg.1 ENST00000444826.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC02000565.hg.1 NR_027714.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC16_KI270728v1_random00006433.hg.1 ENST00000618201.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008431.hg.1 ENST00000413404.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008431.hg.1 ENST00000413404.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008431.hg.1 NR_027714.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200008431.hg.1 NR_027714.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02008643.hg.1 ENST00000444826.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02008643.hg.1 NR_027714.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02008643.hg.1 TC02000565.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0200008431.hg.1 ENST00000413404.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008431.hg.1 ENST00000413404.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008431.hg.1 NR_027714.1 Refseq
          Affymetrix Human Clariom D Assay TC0200008431.hg.1 NR_027714.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200158407.hg.1 ENST00000413404.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200158407.hg.1 NR_027714.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200158407.hg.1 ENST00000413404.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200158407.hg.1 NR_027714.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200158407.hg.1 TC0200008431.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200158408.hg.1 NR_027714.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200158408.hg.1 NR_027714.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200158408.hg.1 TC0200008431.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC16_KI270728v1_random00006433.hg.1 ENST00000618201.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006476.hg.1 ENST00000623630.1 Ensembl
          Human Genome U133 Plus 2.0 Array 224425_x_at - -
          Human Genome U133 Plus 2.0 Array 224425_PM_x_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224425_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224425_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224425_x_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224425_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224425_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224425_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224425_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224425_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224425_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224425_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224425_x_at4 - -
          Human Genome U133 Set 224425_x_at - -
          Human Genome U133 Set 224425_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A015973 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A015973 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 696998 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1769703 NR_027714.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1769703 AY026351.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1769703 NR_027714.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1769703 AY026351.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1769703 NR_027714.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1769703 AY026351.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1769703 NR_027714.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1769703 AY026351.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1769703 NR_027714.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1769703 AY026351.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline