| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010623.hg.1 |
ENST00000440574.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010623.hg.1 |
ENST00000440574.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010623.hg.1 |
ENST00000440574.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010623.hg.1 |
ENST00000448588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010623.hg.1 |
ENST00000448588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010623.hg.1 |
ENST00000448588.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010623.hg.1 |
NR_046452.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010623.hg.1 |
NR_046452.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010623.hg.1 |
NR_046452.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200175243.hg.1 |
ENST00000440574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200175243.hg.1 |
ENST00000448588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200175243.hg.1 |
NR_046452.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200175243.hg.1 |
ENST00000440574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200175243.hg.1 |
ENST00000448588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200175243.hg.1 |
NR_046452.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200175243.hg.1 |
TC0200010623.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200175242.hg.1 |
ENST00000440574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200175242.hg.1 |
NR_046452.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200175242.hg.1 |
ENST00000440574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200175242.hg.1 |
NR_046452.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200175242.hg.1 |
TC0200010623.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200175245.hg.1 |
ENST00000440574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200175245.hg.1 |
ENST00000448588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200175245.hg.1 |
NR_046452.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200175245.hg.1 |
ENST00000440574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200175245.hg.1 |
ENST00000448588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200175245.hg.1 |
NR_046452.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200175245.hg.1 |
TC0200010623.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200010623.hg.1 |
ENST00000440574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010623.hg.1 |
ENST00000440574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010623.hg.1 |
ENST00000440574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010623.hg.1 |
ENST00000448588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010623.hg.1 |
ENST00000448588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010623.hg.1 |
ENST00000448588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010623.hg.1 |
NR_046452.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010623.hg.1 |
NR_046452.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010623.hg.1 |
NR_046452.1 |
Refseq |