Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02341514_g1
          See other OR4Q3 GE Assays ›
          Gene Symbol:
          OR4Q3
          Gene Name:
          olfactory receptor family 4 subfamily Q member 3
          Gene Aliases:
          C14orf13, HSA6, OR14-3, OR4Q4, c14_5008
          Chromosome Location:
          Chr.14: 19747428 - 19748369 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02341514_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR4Q3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172194.1 NP_751944.1 1 757 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400006484.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000034.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OR4Q3

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29608408 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16682357 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16682306 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29608474 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29608458 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09152339 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29608395 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29608432 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29608431 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16682312 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16682301 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09152342 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16682330 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29608453 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29608412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09152337 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16682377 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29608414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29608406 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16682327 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16682356 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09152343 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09152346 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09152341 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3315979 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3262236 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3144 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3145 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 35289 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 59152 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P33468099 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3315979 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3262236 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3315979 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3262236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608414 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682306 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682312 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682310 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09152337 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09152339 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608431 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608408 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682308 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608395 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608407 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682327 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682304 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682330 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682301 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09152341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682298 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09152344 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682327 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682301 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682308 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682310 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608431 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09152344 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608408 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608414 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608395 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682312 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682304 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682298 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09152337 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682306 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16682330 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09152341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09152339 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29608407 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09152337 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09152338 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09152344 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09152341 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09152339 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09152340 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09152336 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P09152335 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09152338 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09152341 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09152348 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09152336 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_172194_at - -
          Affymetrix Human Gene 1.0 ST Array 7973005 - -
          Affymetrix Human Gene 1.1 ST Array 7973004 - -
          Affymetrix Human Genome U219 Array 11742582_at - -
          Affymetrix Human Genome U219 Array 11742582_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1400006484.hg.1 ENST00000331723.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400006484.hg.1 NM_172194.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006496.hg.1 ENST00000557009.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14000150.hg.1 ENST00000331723.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14000150.hg.1 NM_172194.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14000150.hg.1 TC14000034.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1400006484.hg.1 ENST00000331723.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400006484.hg.1 NM_172194.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006496.hg.1 ENST00000557009.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400144303.hg.1 ENST00000331723.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400144303.hg.1 NM_172194.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400144303.hg.1 ENST00000331723.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400144303.hg.1 NM_172194.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400144303.hg.1 TC1400006484.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A051228 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A051228 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2226403 NM_172194.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1737373 AK097598.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1737373 BC062994.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1737373 NR_015416.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1737373 NM_172194.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1737373 AK097598.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1737373 BC062994.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1737373 NR_015416.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1737373 NM_172194.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2226403 NM_172194.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1737373 AK097598.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1737373 BC062994.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1737373 NR_015416.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1737373 NM_172194.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2226403 NM_172194.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2226403 NM_172194.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_9835-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1737373 AK097598.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1737373 BC062994.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1737373 NR_015416.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1737373 NM_172194.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1737373 AK097598.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1737373 BC062994.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1737373 NR_015416.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1737373 NM_172194.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 1660148 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_9835 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2226403 NM_172194.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2226403 NM_172194.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1737373 AK097598.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1737373 BC062994.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1737373 NR_015416.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1737373 NM_172194.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1737373 AK097598.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1737373 BC062994.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1737373 NR_015416.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1737373 NM_172194.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2226403 NM_172194.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1737373 AK097598.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1737373 BC062994.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1737373 NR_015416.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1737373 NM_172194.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2226403 NM_172194.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline