Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02380072_u1
          See other NDUFAF2 GE Assays ›
          Gene Symbol:
          NDUFAF2
          Gene Name:
          NADH:ubiquinone oxidoreductase complex assembly factor 2
          Gene Aliases:
          B17.2L, MMTN, NDUFA12L, mimitin
          Chromosome Location:
          Chr.5: 60945129 - 61153037 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02380072_u1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NDUFAF2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_174889.4 NP_777549.1 4 411 131
          GenBank mRNA AB183433.1 - 4 284 131
          AF087990.1 - 4 380 131
          AK291296.1 - 4 374 131
          BC001753.1 - 1 333 3354845 131
          BC033965.1 - 4 327 5288040 131
          BC070357.1 - 3 289 131
          KU178834.1 - 4 284 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500007542.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05000260.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NDUFAF2

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          respiratory electron transport chain
          mitochondrial respiratory chain complex I assembly
          cellular respiration
          negative regulation of insulin secretion involved in cellular response to glucose stimulus
          reactive oxygen species metabolic process
          NADH dehydrogenase (ubiquinone) activity
          electron carrier activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05003159.hg.1 ENST00000296597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003159.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003159.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05003159.hg.1 TC05000260.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05003160.hg.1 ENST00000296597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003160.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003160.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05003160.hg.1 TC05000260.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100012832.hg.1 ENST00000526902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012832.hg.1 ENST00000526902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003157.hg.1 ENST00000296597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003157.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05003157.hg.1 TC05000260.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05003163.hg.1 ENST00000296597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003163.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003163.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05003163.hg.1 TC05000260.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200011913.hg.1 ENST00000624084.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05003165.hg.1 ENST00000296597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003165.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003165.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003165.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05003165.hg.1 TC05000260.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05003166.hg.1 ENST00000296597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003166.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003166.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05003166.hg.1 NM_174889.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05003166.hg.1 TC05000260.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay TC0500007542.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012832.hg.1 ENST00000526902.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012832.hg.1 ENST00000526902.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150820.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150820.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150820.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150820.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150820.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150820.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150820.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150820.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150820.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150820.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150820.hg.1 TC0500007542.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500150810.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150810.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150810.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150810.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150810.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150810.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150810.hg.1 TC0500007542.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500150811.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150811.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150811.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150811.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150811.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150811.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150811.hg.1 TC0500007542.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500150816.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150816.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150816.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150816.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150816.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150816.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150816.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150816.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150816.hg.1 TC0500007542.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0200011913.hg.1 ENST00000624084.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150807.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150807.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150807.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150807.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150807.hg.1 TC0500007542.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500150812.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150812.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150812.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150812.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150812.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150812.hg.1 ENST00000502658.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150812.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150812.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150812.hg.1 TC0500007542.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500150819.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150819.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150819.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150819.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150819.hg.1 ENST00000296597.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150819.hg.1 ENST00000511107.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150819.hg.1 ENST00000512623.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500150819.hg.1 NM_174889.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500150819.hg.1 TC0500007542.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline