Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02384823_m1
          See other AQP7P3 GE Assays ›
          Gene Symbol:
          AQP7P3
          Gene Name:
          aquaporin 7 pseudogene 3
          Gene Aliases:
          AQPap-3
          Chromosome Location:
          Chr.9: 65877574 - 65912560 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02384823_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AQP7P3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026558.1 2 - 3 122 126
          GenBank mRNA BC052332.1 - 2 - 3 122 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900010101.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000250.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          water transport
          cellular water homeostasis
          glycerol transport
          carbohydrate transmembrane transport
          ion transmembrane transport
          urea transmembrane transport
          urea transmembrane transporter activity
          water channel activity
          glycerol channel activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 933924 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 788202 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 522821 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 757918 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 795960 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 159848 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 336401 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 362992 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 240969 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 497519 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 124398 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 735383 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 113659 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 48446 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 408801 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 28572 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 155654 AK289805.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 155654 BC029500.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 155654 BC041667.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 155654 BX648945.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 155654 NM_001990.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 155654 U81602.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 387947 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 43718 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 164884 AF257472.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 164884 AK022689.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 164884 AY358608.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 164884 NM_024944.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 299856 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 418584 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 2016 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 64968 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 195755 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 282148 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 148109 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 50819 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 387266 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 94659 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 147760 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 394756 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 92306 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 40159 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3371115 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4671 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4672 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 56484 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3371115 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3371115 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09002815.hg.1 TC09000250.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09002815.hg.1 NR_026558.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09002814.hg.1 TC09000250.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09002814.hg.1 NR_026558.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09002816.hg.1 TC09000250.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09002816.hg.1 NR_026558.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09002813.hg.1 TC09000250.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09002813.hg.1 NR_026558.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900169405.hg.1 NR_026558.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900169405.hg.1 NR_026558.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900169405.hg.1 TC0900010101.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900169406.hg.1 NR_026558.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900169406.hg.1 NR_026558.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900169406.hg.1 TC0900010101.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900169419.hg.1 NR_026558.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900169419.hg.1 NR_026558.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900169419.hg.1 TC0900010101.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037492 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038629 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037492 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038629 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0