Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02387400_g1
          See other NANOG GE Assays ›
          Gene Symbol:
          NANOG
          Gene Name:
          Nanog homeobox
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 7789396 - 7796061 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02387400_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NANOG
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001297698.1 NP_001284627.1 1 - 2 366 109
          NM_024865.3 NP_079141.2 1 - 2 366 109
          XM_011520850.1 XP_011519152.1 1 - 2 366 109
          GenBank mRNA AB093576.1 - 1 - 2 363 109
          AK022643.1 - 1 - 2 363 109
          AK290896.1 - 1 - 2 332 109
          AY230262.1 - 1 - 2 147 109
          AY578089.1 - 1 - 2 147 109
          DC425336.1 - 1 - 2 367 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200006688.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000114.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Homo sapiens Human Stem Cell Pluripotency Human Transcriptional Regulatory Network in Embryonic Stem C 1700 Inventoried Human_Stem_Cell Human_Transcriptional_Regulatory_Network_in_Embryonic_Stem_Cell Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NANOG

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          endodermal cell fate specification
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cell proliferation
          regulation of gene expression
          stem cell population maintenance
          cell differentiation
          somatic stem cell population maintenance
          regulation of cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of nucleic acid-templated transcription
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          protein binding
          sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1500009012.hg.1 ENST00000528386.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009012.hg.1 ENST00000528386.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009012.hg.1 ENST00000528386.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009012.hg.1 ENST00000629403.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009012.hg.1 ENST00000629403.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008228.hg.1 ENST00000424456.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00009905.hg.1 ENST00000439890.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000526434.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000526434.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000526434.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12001584.hg.1 ENST00000229307.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12001584.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12001584.hg.1 NM_024865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12001584.hg.1 TC12000114.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0X00009481.hg.1 ENST00000453325.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011593.hg.1 ENST00000429190.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007044.hg.1 ENST00000427230.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009987.hg.1 ENST00000554450.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12001582.hg.1 ENST00000229307.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12001582.hg.1 NM_024865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12001582.hg.1 TC12000114.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12001585.hg.1 ENST00000229307.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12001585.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12001585.hg.1 NM_024865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12001585.hg.1 TC12000114.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1400009987.hg.1 ENST00000554450.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146794.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146794.hg.1 ENST00000526434.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146794.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200146794.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200146794.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146794.hg.1 ENST00000526434.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146794.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200146794.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200146794.hg.1 TC1200006688.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000526434.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000526434.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000526434.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay TC1200006688.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00009481.hg.1 ENST00000453325.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146793.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146793.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200146793.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200146793.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146793.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200146793.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200146793.hg.1 TC1200006688.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200146797.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146797.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146797.hg.1 ENST00000526434.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146797.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146797.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200146797.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200146797.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146797.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146797.hg.1 ENST00000526434.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146797.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146797.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200146797.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200146797.hg.1 TC1200006688.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200146798.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146798.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146798.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146798.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200146798.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200146798.hg.1 ENST00000229307.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146798.hg.1 ENST00000526286.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146798.hg.1 ENST00000541267.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200146798.hg.1 NM_001297698.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200146798.hg.1 NM_024865.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200146798.hg.1 TC1200006688.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700007044.hg.1 ENST00000427230.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008228.hg.1 ENST00000424456.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00009905.hg.1 ENST00000439890.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009012.hg.1 ENST00000528386.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009012.hg.1 ENST00000528386.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009012.hg.1 ENST00000528386.3 Ensembl
          Affymetrix Human Clariom D Assay TC1500009012.hg.1 ENST00000629403.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009012.hg.1 ENST00000629403.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011593.hg.1 ENST00000429190.2 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 185136 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline