Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02576269_s1
          See other ADAMTSL4-AS1 GE Assays ›
          Gene Symbol:
          ADAMTSL4-AS1
          Gene Name:
          ADAMTSL4 antisense RNA 1
          Gene Aliases:
          C1orf138
          Chromosome Location:
          Chr.1: 150560895 - 150574552 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02576269_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ADAMTSL4-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_104133.1 4 776 66
          GenBank mRNA AK127688.1 - 1 961 66
          BC132990.1 - 1 339 66
          BC132992.1 - 1 339 66
          BG231546.1 - 1 270 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100018485.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 47904 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7919746 - -
          Affymetrix Human Gene 1.0 ST Array 7919744 - -
          Affymetrix Human Gene 1.0 ST Array 7919745 - -
          Affymetrix Human Gene 1.1 ST Array 7919743 - -
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 ENST00000369035.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 ENST00000369035.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 ENST00000617352.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 ENST00000617352.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 ENST00000617352.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 ENST00000617352.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 ENST00000617352.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 NR_104133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 NR_104133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 NR_104133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 NR_104133.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018485.hg.1 NR_104133.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 ENST00000369035.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 ENST00000369035.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 ENST00000617352.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 ENST00000617352.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 ENST00000617352.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 ENST00000617352.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 ENST00000617352.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 NR_104133.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 NR_104133.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 NR_104133.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 NR_104133.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018485.hg.1 NR_104133.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100243083.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243083.hg.1 ENST00000617352.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243083.hg.1 NR_104133.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100243083.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243083.hg.1 ENST00000617352.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243083.hg.1 NR_104133.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100243083.hg.1 TC0100018485.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100243085.hg.1 ENST00000369035.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243085.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243085.hg.1 ENST00000369035.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243085.hg.1 ENST00000615012.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243085.hg.1 TC0100018485.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1705904 BC132990.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1705904 BC132992.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1705904 AK127688.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1705904 BC132990.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1705904 BC132992.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1705904 AK127688.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1705904 BC132990.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1705904 BC132992.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1705904 AK127688.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1705904 BC132990.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1705904 BC132992.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1705904 AK127688.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1705904 BC132990.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1705904 BC132992.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1705904 AK127688.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1705904 BC132990.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1705904 BC132992.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1705904 AK127688.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1705904 BC132990.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1705904 BC132992.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1705904 AK127688.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline