Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02576480_m1
          See other CD34 GE Assays ›
          Gene Symbol:
          CD34
          Gene Name:
          CD34 molecule
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 207886538 - 207911338 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02576480_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CD34
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025109.1 NP_001020280.1 4 - 5 860 63
          NM_001773.2 NP_001764.1 4 - 5 860 63
          GenBank mRNA AF202879.1 - 2 - 3 105 63
          AF523361.1 - 4 - 5 673 63
          AF538601.1 - 3 - 4 511 63
          AK291529.1 - 4 - 5 613 63
          AK293030.1 - 4 - 5 637 63
          AK294381.1 - 3 - 4 384 63
          AK300835.1 - 5 - 6 677 63
          BC039146.1 - 4 - 5 610 4746591 63
          M81104.1 - 4 - 5 859 63
          S53910.1 - 4 - 5 692 63
          S53911.1 - 4 - 5 692 63

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CD34

          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          tissue homeostasis
          endothelial cell proliferation
          glomerular filtration
          endothelium development
          cell-matrix adhesion
          signal transduction
          regulation of blood pressure
          cell proliferation
          positive regulation of gene expression
          negative regulation of gene expression
          single organismal cell-cell adhesion
          hemopoiesis
          negative regulation of blood coagulation
          negative regulation of tumor necrosis factor production
          positive regulation of interleukin-10 production
          mesangial cell-matrix adhesion
          paracrine signaling
          positive regulation of odontogenesis
          negative regulation of nitric oxide biosynthetic process
          positive regulation of angiogenesis
          cell motility
          regulation of immune response
          leukocyte migration
          transdifferentiation
          vascular wound healing
          hematopoietic stem cell proliferation
          positive regulation of transforming growth factor beta production
          positive regulation of granulocyte colony-stimulating factor production
          extracellular exosome assembly
          glomerular endothelium development
          stem cell proliferation
          metanephric glomerular mesangial cell differentiation
          negative regulation of cellular response to heat
          negative regulation of cellular response to hypoxia
          negative regulation of interleukin-2 secretion
          positive regulation of glial cell-derived neurotrophic factor secretion
          negative regulation of neuron death
          positive regulation of vasculogenesis
          transcription factor binding
          carbohydrate binding
          sulfate binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000310833.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000356522.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000367036.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 ENST00000485761.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001025109.1 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq
          Affymetrix Human Clariom D Assay TC0100017142.hg.1 NM_001773.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0