Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02598440_g1
          See other POTEKP GE Assays ›
          Gene Symbol:
          POTEKP
          Gene Name:
          POTE ankyrin domain family member K, pseudogene
          Gene Aliases:
          ACT, ACTBL3, POTE2delta, POTEK
          Chromosome Location:
          Chr.2: 131592093 - 131609401 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02598440_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: POTEKP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AY014272.1 - 1 838 136

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21602081 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21602087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15328821 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21602086 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 5690 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 20309 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P155776 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P825874 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7973000 - -
          Affymetrix Human Gene 1.1 ST Array 8045321 - -
          Affymetrix Human Gene 1.1 ST Array 8045208 - -
          Affymetrix Human Gene 1.1 ST Array 7977454 - -
          Affymetrix HT HG-U133+ PM Array Plate 210926_PM_at - -
          Human Genome U133 Plus 2.0 Array 210926_PM_at - -
          Human Genome U133 Plus 2.0 Array 210926_at - -
          Human Genome U133A 2.0 Array 210926_at - -
          Human Genome U133A 2.0 Array 210926_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw594415 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210926_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210926_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210926_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210926_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210926_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210926_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210926_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210926_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210926_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210926_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210926_at9 - -
          Human Genome U133 Set 210926_at - -
          Human Genome U133 Set 210926_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A016376 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A016376 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1814998 AY014272.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1814998 NR_033885.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2112673 AY014272.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2112673 NR_033885.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1814998 AY014272.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1814998 NR_033885.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1814998 AY014272.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1814998 NR_033885.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2112673 AY014272.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2112673 NR_033885.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1814998 AY014272.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1814998 NR_033885.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2112673 AY014272.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2112673 NR_033885.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2112673 AY014272.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2112673 NR_033885.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1814998 AY014272.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1814998 NR_033885.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2112673 AY014272.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2112673 NR_033885.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline