Search
Search
| Gene Symbol: GLDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000170.2 | NP_000161.2 | 10 - 11 | 1588 | 94 | |
| GenBank mRNA | AK310534.1 | - | 6 - 7 | 963 | 94 | |
| AK314156.1 | - | 10 - 11 | 1590 | 94 | ||
| BC111993.1 | - | 10 - 11 | 1461 | 8327461 | 94 | |
| BC111995.1 | - | 10 - 11 | 1461 | 8327463 | 94 | |
| D90239.1 | - | 10 - 11 | 1545 | 94 | ||
| M63635.1 | - | 10 - 11 | 1476 | 94 | ||
| M64590.1 | - | 10 - 11 | 1545 | 94 | ||
Set Membership: |
|