Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02787889_g1
          See other CCL2 GE Assays ›
          Gene Symbol:
          CCL2
          Gene Name:
          C-C motif chemokine ligand 2
          Gene Aliases:
          GDCF-2, HC11, HSMCR30, MCAF, MCP-1, MCP1, SCYA2, SMC-CF
          Chromosome Location:
          Chr.17: 34255285 - 34257203 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02787889_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCL2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002982.4 NP_002973.1 2 - 3 255 62
          GenBank mRNA AK311960.1 - 2 - 3 255 62
          AV733621.2 - 2 - 3 263 62
          BC009716.1 - 2 - 3 246 62
          BG530064.1 - 2 - 3 276 62
          M24545.1 - 2 - 3 259 62
          M28226.1 - 2 - 3 236 62
          S69738.1 - 2 - 3 221 62
          S71513.1 - 2 - 3 243 62
          X14768.1 - 2 - 3 243 62

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCL2

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          monocyte chemotaxis
          protein phosphorylation
          chemotaxis
          inflammatory response
          immune response
          humoral immune response
          cytoskeleton organization
          cell adhesion
          signal transduction
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          JAK-STAT cascade
          regulation of cell shape
          response to bacterium
          organ morphogenesis
          positive regulation of gene expression
          positive regulation of macrophage chemotaxis
          viral genome replication
          cytokine-mediated signaling pathway
          sensory perception of pain
          cellular homeostasis
          positive regulation of cell migration
          negative regulation of glial cell apoptotic process
          helper T cell extravasation
          chemokine (C-C motif) ligand 2 signaling pathway
          negative regulation of neuron apoptotic process
          astrocyte cell migration
          cellular response to fibroblast growth factor stimulus
          eosinophil chemotaxis
          macrophage chemotaxis
          positive regulation of T cell activation
          positive chemotaxis
          positive regulation of synaptic transmission, glutamatergic
          antimicrobial humoral immune response mediated by antimicrobial peptide
          chemokine-mediated signaling pathway
          cellular response to lipopolysaccharide
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          positive regulation of calcium ion import
          positive regulation of glutamate receptor signaling pathway
          negative regulation of vascular endothelial cell proliferation
          negative regulation of G1/S transition of mitotic cell cycle
          positive regulation of endothelial cell apoptotic process
          positive regulation of apoptotic cell clearance
          negative regulation of natural killer cell chemotaxis
          protein kinase activity
          receptor binding
          cytokine activity
          protein binding
          chemokine activity
          CCR2 chemokine receptor binding
          chemoattractant activity
          CCR chemokine receptor binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline