Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02800695_m1
          See other HPRT1 GE Assays ›
          Gene Symbol:
          HPRT1
          Gene Name:
          hypoxanthine phosphoribosyltransferase 1
          Gene Aliases:
          HGPRT, HPRT
          Chromosome Location:
          Chr.X: 134460145 - 134500668 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02800695_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HPRT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000194.2 NP_000185.1 2 - 3 297 82
          GenBank mRNA AK313435.1 - 2 - 3 277 82
          BC000578.2 - 2 - 3 169 3163726 82
          BT019350.1 - 2 - 3 130 82
          CR407645.1 - 2 - 3 130 82
          L29382.1 - 2 - 3 130 82
          M24772.1 - 114 82
          M31642.1 - 2 - 3 215 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00011326.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000636.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Endogenous Controls Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HPRT1

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to amphetamine
          purine nucleotide biosynthetic process
          purine ribonucleoside salvage
          adenine salvage
          guanine salvage
          grooming behavior
          locomotory behavior
          cytolysis
          striatum development
          cerebral cortex neuron differentiation
          central nervous system neuron development
          GMP salvage
          IMP salvage
          dopamine metabolic process
          purine-containing compound salvage
          hypoxanthine salvage
          positive regulation of dopamine metabolic process
          GMP catabolic process
          IMP metabolic process
          hypoxanthine metabolic process
          lymphocyte proliferation
          dendrite morphogenesis
          protein homotetramerization
          nucleotide binding
          magnesium ion binding
          hypoxanthine phosphoribosyltransferase activity
          protein binding
          protein homodimerization activity
          guanine phosphoribosyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X008364.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X008364.hg.1 ENST00000370796.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X008364.hg.1 ENST00000462974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X008364.hg.1 NM_000194.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X008364.hg.1 TC0X000636.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0X00177946.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00177946.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00177946.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00177946.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00177946.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00177946.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00177946.hg.1 TC0X00011326.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000298556.7 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 ENST00000462974.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011326.hg.1 NM_000194.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-hqqtp:80/100.66.76.16:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline