Search
Search

G6PC1| Gene Symbol: G6PC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000151.4 | NP_000142.2 | 4 - 5 | 644 | 77 | |
| NM_001270397.2 | NP_001257326.1 | 4 - 5 | 567 | 77 | ||
| GenBank mRNA | AK303771.1 | - | 4 - 5 | 569 | 77 | |
| AK313982.1 | - | 4 - 5 | 641 | 77 | ||
| BC130478.1 | - | 4 - 5 | 612 | 77 | ||
| BC136369.1 | - | 4 - 5 | 612 | 77 | ||
| U01120.1 | - | 4 - 5 | 77 | |||
Set Membership: |
|