Search
Search
| Gene Symbol: GLO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006708.2 | NP_006699.2 | 3 - 4 | 427 | 90 | |
| GenBank mRNA | AB209801.1 | - | 3 - 4 | 318 | 90 | |
| AK129573.1 | - | 3 - 4 | 360 | 90 | ||
| AK130312.1 | - | 3 - 4 | 324 | 90 | ||
| AK312662.1 | - | 3 - 4 | 396 | 90 | ||
| AL833442.1 | - | 3 - 4 | 377 | 90 | ||
| BC001741.1 | - | 3 - 4 | 368 | 3352922 | 90 | |
| BC011365.1 | - | 3 - 4 | 381 | 4182593 | 90 | |
| BC015934.1 | - | 3 - 4 | 352 | 3912185 | 90 | |
| BT019987.1 | - | 3 - 4 | 305 | 90 | ||
| BT019988.1 | - | 3 - 4 | 305 | 90 | ||
| D13315.1 | - | 3 - 4 | 392 | 90 | ||
| DA342581.1 | - | 3 - 4 | 427 | 90 | ||
| FJ224325.1 | - | 3 - 4 | 392 | 90 | ||
| KJ534846.1 | - | 3 - 4 | 305 | 90 | ||
| L07837.1 | - | 3 - 4 | 365 | 90 | ||
| S83285.1 | - | 3 - 4 | 314 | 90 | ||
Set Membership: |
|