Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03043809_m1
          See other SIRPG GE Assays ›
          Gene Symbol:
          SIRPG
          Gene Name:
          signal regulatory protein gamma
          Gene Aliases:
          CD172g, SIRP-B2, SIRPB2, SIRPgamma, bA77C3.1
          Chromosome Location:
          Chr.20: 1629152 - 1686516 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03043809_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SIRPG
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039508.1 NP_001034597.1 4 - 5 896 96
          NM_018556.3 NP_061026.2 5 - 6 1229 96
          NM_080816.2 NP_543006.2 3 - 4 578 96
          XM_005260749.3 XP_005260806.1 4 - 5 947 96
          XM_011529286.2 XP_011527588.1 5 - 6 1276 96
          GenBank mRNA AB042624.1 - 5 - 6 1245 96
          AK292745.1 - 5 - 6 1229 96
          BC020629.1 - 3 - 4 555 96
          BC064532.1 - 5 - 6 1190 5588038 96
          DB046293.1 - 2 - 3 262 96

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SIRPG

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          cell-cell signaling
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          positive regulation of cell-cell adhesion
          intracellular signal transduction
          positive regulation of T cell activation
          leukocyte migration
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000216927.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000216927.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000216927.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000216927.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000344103.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000344103.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000344103.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000344103.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000344103.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000344103.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000478145.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000478145.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000478145.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000478145.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000497407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000497407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 ENST00000497407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_080816.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_080816.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_080816.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_080816.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_080816.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009971.hg.1 NM_080816.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000216927.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000216927.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000216927.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000216927.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000303415.7 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000344103.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000344103.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000344103.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000344103.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000344103.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000344103.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381580.5 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000381583.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000478145.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000478145.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000478145.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000478145.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000497407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000497407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 ENST00000497407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_001039508.1 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_018556.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_080816.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_080816.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_080816.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_080816.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_080816.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009971.hg.1 NM_080816.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 206705 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline