Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03043962_g1
          See other KLHDC3 GE Assays ›
          Gene Symbol:
          KLHDC3
          Gene Name:
          kelch domain containing 3
          Gene Aliases:
          PEAS, dJ20C7.3
          Chromosome Location:
          Chr.6: 43014103 - 43021298 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03043962_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KLHDC3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057161.3 NP_476502.1 5 - 6 821 86
          NR_040101.1 4 - 5 705 86
          GenBank mRNA AB055925.1 - 5 - 6 685 86
          AK097719.1 - 3 - 4 547 86
          AK290384.1 - 5 - 6 677 86
          AK302336.1 - 4 - 5 460 86
          AK308917.1 - 5 - 6 707 86
          BC000295.2 - 5 - 6 641 2819570 86
          BC001789.1 - 5 - 6 656 3355470 86
          BC001793.2 - 5 - 6 619 3352397 86
          BC007296.1 - 5 - 6 671 3349731 86
          BC009460.1 - 5 - 6 669 3537223 86
          BC021546.1 - 5 - 6 658 3351635 86
          BC041793.1 - 4 - 5 559 5288540 86
          BC045612.1 - 4 - 5 559 5313255 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600008066.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000592.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KLHDC3

          Gene Ontology Categories:

          Function(s) Process(es)

          reciprocal meiotic recombination
          IRE1-mediated unfolded protein response
          chromatin binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06006789.hg.1 ENST00000244670.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006789.hg.1 ENST00000326974.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006789.hg.1 ENST00000332245.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006789.hg.1 ENST00000394096.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006789.hg.1 ENST00000426116.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006789.hg.1 ENST00000432243.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06006789.hg.1 NM_057161.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06006789.hg.1 NR_040101.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06006789.hg.1 TC06000592.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay TC0600008066.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600156087.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0600156087.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600156087.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600156087.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600156087.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600156087.hg.1 ENST00000244670.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0600156087.hg.1 ENST00000326974.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600156087.hg.1 ENST00000332245.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600156087.hg.1 NM_057161.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600156087.hg.1 NR_040101.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600156087.hg.1 TC0600008066.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline