Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03044163_g1
          See other RPL35 GE Assays ›
          Gene Symbol:
          RPL35
          Gene Name:
          ribosomal protein L35
          Gene Aliases:
          L35
          Chromosome Location:
          Chr.9: 124857879 - 124861961 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03044163_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RPL35
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007209.3 NP_009140.1 2 - 3 182 117
          GenBank mRNA AK291072.1 - 2 - 3 176 117
          BC000348.2 - 2 - 3 166 2960987 117
          BC010919.1 - 1 182 4251487 117
          BC071915.1 - 1 148 4896387 117
          BC094828.1 - 1 136 4548321 117
          BM854016.1 - 2 - 3 182 117
          CR456877.1 - 1 - 2 133 117
          CR996819.1 - 2 - 3 167 117
          U12465.1 - 2 - 3 160 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900011510.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RPL35

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          maturation of LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          rRNA processing
          translation
          translational initiation
          SRP-dependent cotranslational protein targeting to membrane
          viral transcription
          mRNA binding
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000348462.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000348462.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000348462.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000348462.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000495728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000495728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000629845.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000629845.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 ENST00000629845.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 NM_007209.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 NM_007209.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 NM_007209.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011510.hg.1 NM_007209.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000402419.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000402419.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006619.hg.1 ENST00000588562.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006619.hg.1 ENST00000588562.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000348462.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000348462.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000348462.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000348462.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000495728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000495728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000629845.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000629845.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 ENST00000629845.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 NM_007209.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 NM_007209.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 NM_007209.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011510.hg.1 NM_007209.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900178470.hg.1 ENST00000348462.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178470.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178470.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178470.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178470.hg.1 NM_007209.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900178470.hg.1 ENST00000348462.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178470.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178470.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178470.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178470.hg.1 NM_007209.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900178470.hg.1 TC0900011510.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1900006619.hg.1 ENST00000588562.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006619.hg.1 ENST00000588562.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000402419.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000402419.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 ENST00000607016.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Clariom D Assay TC0600014283.hg.1 NM_006703.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900178474.hg.1 ENST00000348462.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178474.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178474.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178474.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178474.hg.1 ENST00000629845.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178474.hg.1 NM_007209.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900178474.hg.1 ENST00000348462.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178474.hg.1 ENST00000373570.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178474.hg.1 ENST00000487431.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178474.hg.1 ENST00000493018.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178474.hg.1 ENST00000629845.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900178474.hg.1 NM_007209.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900178474.hg.1 TC0900011510.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline