| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016806.hg.1 |
ENST00000300773.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016806.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016806.hg.1 |
TC11001333.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000300773.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000300773.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NR_038321.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NR_038321.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NR_038321.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NR_038321.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NR_038321.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013135.hg.1 |
NR_038321.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016807.hg.1 |
ENST00000300773.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016807.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016807.hg.1 |
TC11001333.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100203422.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203422.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203422.hg.1 |
TC1100013135.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000300773.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000300773.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000415970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000418729.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
ENST00000420726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NR_038321.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NR_038321.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NR_038321.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NR_038321.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NR_038321.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013135.hg.1 |
NR_038321.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203423.hg.1 |
ENST00000300773.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203423.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203423.hg.1 |
ENST00000300773.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203423.hg.1 |
NM_001005567.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203423.hg.1 |
TC1100013135.hg.1 |
Clariom D TC |