Search
Search
| Gene Symbol: CSH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020991.3 | NP_066271.1 | 5 | 656 | 153 | |
| NM_022644.3 | NP_072170.1 | 4 | 909 | 153 | ||
| NM_022645.2 | NP_072171.1 | 3 | 371 | 153 | ||
| GenBank mRNA | BC022044.1 | - | 5 | 607 | 4769046 | 153 |
| BC035965.1 | - | 5 | 603 | 4731880 | 153 | |
| BC119748.2 | - | 5 | 600 | 40119242 | 153 | |
| V00573.1 | - | 5 | 565 | 153 | ||
Set Membership: |
|