Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03297478_s1
          See other TPI1P2 GE Assays ›
          Gene Symbol:
          TPI1P2
          Gene Name:
          triosephosphate isomerase 1 pseudogene 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 129055223 - 129057239 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03297478_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TPI1P2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002187.3 1 751 111
          GenBank mRNA AK094894.1 - 1 753 111
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI001983.1 - 1 61 Chr. 7 - 129055915 - 129056401 [+] on Build GRCh38 110

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transportin 3 TNPO3 IPO12,
          LGMD1F,
          MTR10A,
          TRN-SR,
          TRN-SR2,
          TRNSR
          -50.0 Anti-sense Chr. 7 - 128954180 - 129055173 on NCBI Build GRCh38
          Downstream mitogen-activated protein kinase kinase 2 pseudogene LOC407835 69.0kb Sense Chr. 7 - 129126271 - 129128007 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700009075.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000790.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK094894.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05776044 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26033763 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26033768 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05776037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05776038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05776042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05776040 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05776039 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 48674 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P358328 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P358328 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 36237 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 43790 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P358328 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P358328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776029 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16017290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033742 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776024 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776006 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033733 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033735 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033758 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033760 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033756 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776009 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033755 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776029 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033748 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16017290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033742 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033733 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776015 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776006 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776024 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033760 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033753 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033735 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033758 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26033743 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05776023 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P358328 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P358328 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01805609 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18112961 - -
          Agilent Human 1B Microarray G4111A A_32_P758502 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776030 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776018 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776021 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776039 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776036 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776014 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776028 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776031 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776023 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776029 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776025 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776020 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776005 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776006 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776040 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776033 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776012 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776013 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776009 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776043 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05776027 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18112961 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002187_at - -
          Affymetrix Human Gene 1.0 ST Array 8136062 - -
          Affymetrix Human Gene 1.1 ST Array 8136061 - -
          Affymetrix Human Transcriptome Array 2.0 PSR07012501.hg.1 NR_002187.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07012501.hg.1 TC07000790.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700009075.hg.1 ENST00000491343.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009075.hg.1 NR_002187.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009075.hg.1 NR_002187.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009075.hg.1 NR_002187.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700164594.hg.1 ENST00000491343.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700164594.hg.1 NR_002187.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700164594.hg.1 ENST00000491343.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700164594.hg.1 NR_002187.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700164594.hg.1 TC0700009075.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700009075.hg.1 ENST00000491343.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700009075.hg.1 NR_002187.3 Refseq
          Affymetrix Human Clariom D Assay TC0700009075.hg.1 NR_002187.3 Refseq
          Affymetrix Human Clariom D Assay TC0700009075.hg.1 NR_002187.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700164593.hg.1 NR_002187.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700164593.hg.1 NR_002187.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700164593.hg.1 TC0700009075.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700164595.hg.1 NR_002187.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700164595.hg.1 NR_002187.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700164595.hg.1 TC0700009075.hg.1 Clariom D TC
          Human Genome U95 Set 78921_PM_i_at - -
          Human Genome U95 Set 78921_i_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035047 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035048 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035047 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035048 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 236495 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1684114 NR_002187.3 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1684114 AA905085.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1684114 AK094894.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1684114 BC053624.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1684114 NR_002187.3 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1684114 AA905085.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1684114 AK094894.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1684114 BC053624.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1684114 NR_002187.3 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1684114 AA905085.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1684114 AK094894.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1684114 BC053624.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1684114 NR_002187.3 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1684114 AA905085.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1684114 AK094894.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1684114 BC053624.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_16558 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_16558 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1684114 NR_002187.3 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1684114 AA905085.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1684114 AK094894.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1684114 BC053624.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1684114 NR_002187.3 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1684114 AA905085.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1684114 AK094894.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1684114 BC053624.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1684114 NR_002187.3 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1684114 AA905085.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1684114 AK094894.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1684114 BC053624.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline