Search
Search

PPP1R2P9| Gene Symbol: PPP1R2P9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002191.2 | 1 | 648 | 125 | ||
| GenBank mRNA | AB044137.1 | - | 1 | 648 | 125 | |
| BC056673.1 | - | 1 | 578 | 125 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calcium/calmodulin dependent serine protein kinase | CASK |
CAGH39, CAMGUK, CMG, FGS4, LIN2, MICPCH, MRXSNA, TNRC8 |
-854.3kb | Sense | Chr. X - 41514934 - 41923525 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927501 | LOC101927501 | 398.8kb | Anti-sense | Chr. X - 43176994 - 43226598 on NCBI Build GRCh38 |
Set Membership: |
|