Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03298719_s1
          See other SCARNA8 GE Assays ›
          Gene Symbol:
          SCARNA8
          Gene Name:
          small Cajal body-specific RNA 8
          Gene Aliases:
          U92
          Chromosome Location:
          Chr.9: 19063656 - 19063786 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03298719_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 4,565.00
          Your Price
          Online offer:
          4565.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SCARNA8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003009.1 1 62 68
          GenBank mRNA AY077742.1 - 1 62 68
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CV569582.1 - 1 258 Chr. 9 - 19063400 - 19063982 [-] on Build GRCh38 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream HAUS augmin like complex subunit 6 HAUS6 Dgt6,
          FAM29A
          Sense Chr. 9 - 19053141 - 19102904 on NCBI Build GRCh38
          Downstream perilipin 2 PLIN2 ADFP,
          ADRP
          44.6kb Sense Chr. 9 - 19108388 - 19127492 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900009646.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000934.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s)

          RNA processing

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4033 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915912 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915944 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915915 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915945 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628011 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915939 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915936 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628005 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915932 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915947 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915929 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915923 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915940 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915928 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915914 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628014 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915922 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915937 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915954 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915912 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915922 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915936 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915932 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915937 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915939 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915945 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915915 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628005 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915954 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915914 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915928 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915947 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915929 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915923 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628014 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915944 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628011 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915940 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06628012 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26915931 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628017 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628019 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628013 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16163520 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628038 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628043 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628028 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628041 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16163522 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628025 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628033 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628030 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16163517 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628005 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628052 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628006 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628021 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628010 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628049 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628045 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628008 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628037 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628047 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628050 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628039 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628015 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06628011 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_003009_at - -
          Affymetrix Human Gene 1.0 ST Array 8160296 - -
          Affymetrix Human Gene 1.1 ST Array 8160295 - -
          Affymetrix Human Transcriptome Array 2.0 TC0900009646.hg.1 ENST00000515924.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009646.hg.1 NR_003009.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09012497.hg.1 ENST00000515924.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09012497.hg.1 NR_003009.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09012497.hg.1 TC09000934.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0900009646.hg.1 ENST00000515924.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009646.hg.1 NR_003009.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900166067.hg.1 ENST00000515924.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900166067.hg.1 NR_003009.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900166067.hg.1 ENST00000515924.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900166067.hg.1 NR_003009.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900166067.hg.1 TC0900009646.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037151 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037151 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3247159 NR_003009.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3247159 AY077742.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3247159 NR_003009.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3247159 AY077742.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3247159 NR_003009.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3247159 AY077742.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3247159 NR_003009.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3247159 AY077742.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline