Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03454013_s1
          See other PCDHB18P GE Assays ›
          Gene Symbol:
          PCDHB18P
          Gene Name:
          protocadherin beta 18 pseudogene
          Gene Aliases:
          PCDH-psi2, PCDHB18
          Chromosome Location:
          Chr.5: 141234333 - 141237529 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03454013_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PCDHB18P
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_001281.2
          1 371 62
          GenBank mRNA AF152528.1 - LIT2076
          PCDH-psi2
          1 153 62
          AK095496.1 -
          1 340 62
          DA121078.1 -
          1 372 62
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA529211.1 - 1 371 Chr. 5 - 141234333 - 141234933 [+] on Build GRCh38 62
          GenBankmRNA DA125678.1 - 1 352 Chr. 5 - 141234352 - 141234868 [+] on Build GRCh38 62
          GenBankmRNA DA483092.1 - 1 340 Chr. 5 - 141234366 - 141235129 [+] on Build GRCh38 62
          GenBankmRNA DA777160.1 - 517 Chr. - - [-] on Build 62
          GenBankmRNA DA186145.1 - 1 371 Chr. 5 - 141234334 - 141234873 [+] on Build GRCh38 62
          GenBankmRNA DB175121.1 - 1 370 Chr. 5 - 141234335 - 141234900 [+] on Build GRCh38 62
          GenBankmRNA DB240188.1 - 1 354 Chr. 5 - 141234352 - 141234922 [+] on Build GRCh38 62
          GenBankmRNA DB099748.1 - 1 369 Chr. 5 - 141234335 - 141234877 [+] on Build GRCh38 62

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protocadherin beta 14 PCDHB14 PCDH-BETA14 -8.3kb Sense Chr. 5 - 141222326 - 141226288 on NCBI Build GRCh38
          Downstream protocadherin beta 19 pseudogene PCDHB19P PCDH-PSI5,
          PCDHB19
          3.2kb Sense Chr. 5 - 141240121 - 141244752 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500008886.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05000758.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK095496.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          calcium ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05010854.hg.1 TC05000758.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05010854.hg.1 ENST00000526308.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05010854.hg.1 NR_001281.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000622990.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000622990.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 ENST00000622990.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161841.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161841.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161841.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161841.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161841.hg.1 TC0500008886.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500161842.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161842.hg.1 ENST00000622990.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161842.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161842.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161842.hg.1 ENST00000622990.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161842.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161842.hg.1 TC0500008886.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500161844.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161844.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161844.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161844.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161844.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161844.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161844.hg.1 TC0500008886.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500161846.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161846.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161846.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161846.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161846.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161846.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161846.hg.1 TC0500008886.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500161845.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161845.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161845.hg.1 ENST00000622990.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161845.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161845.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161845.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161845.hg.1 ENST00000622990.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161845.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161845.hg.1 TC0500008886.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000622990.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000622990.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 ENST00000622990.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008886.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161843.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161843.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161843.hg.1 ENST00000622990.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161843.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161843.hg.1 ENST00000524813.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161843.hg.1 ENST00000526308.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161843.hg.1 ENST00000622990.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500161843.hg.1 NR_001281.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500161843.hg.1 TC0500008886.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline