Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03459205_s1
          See other RBMY2FP GE Assays ›
          Gene Symbol:
          RBMY2FP
          Gene Name:
          RNA binding motif protein, Y-linked, family 2, member F pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.Y: 22308859 - 22316205 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03459205_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 4,565.00
          Your Price
          Online offer:
          4565.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RBMY2FP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002193.2 4 895 146
          GenBank mRNA BC026077.2 - 4 897 146

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream testis-specific transcript,
          Y-linked 5 (non-protein coding)
          TTTY5 LINC00126,
          NCRNA00126,
          TTY5
          -15.6kb Anti-sense Chr. Y - 22296798 - 22298876 on NCBI Build GRCh38
          Downstream RNA binding motif protein,
          Y-linked,
          family 1,
          member A1 pseudogene
          LOC100652931 473.0 Sense Chr. Y - 22316678 - 22320384 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 61354 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 870254 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 359181 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 346420 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 208887 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 223849 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 304228 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 34980 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 194967 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 274264 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 152456 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 313591 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 23682 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 346973 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 210031 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 22853 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 191807 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P424689 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P424689 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 6544 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 26811 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03807739 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P424689 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P424689 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847797 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002349 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847793 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12067216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847810 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847819 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12067213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847813 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002336 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847794 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847799 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847800 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847826 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847804 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847827 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12067214 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847803 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002342 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12067220 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002333 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847808 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847815 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002331 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002348 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12067217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847822 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847818 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002348 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002333 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847822 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002349 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847794 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002336 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12067213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12067214 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847800 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847818 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002342 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847826 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847815 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847799 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847803 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847810 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847808 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002331 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847793 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12067220 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12067216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P12067217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847813 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847819 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847804 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847827 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15002328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32847797 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P424689 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P424689 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002334 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002320 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002352 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002336 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12067207 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002312 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002307 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12067219 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12067216 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002339 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17466520 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002310 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002346 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002326 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12067206 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17466523 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17466530 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12067211 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002347 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002350 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12067215 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P12067212 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002333 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002328 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17466524 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002353 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002341 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002316 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002305 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15002322 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P21675426 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03807739 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002193_at - -
          Affymetrix Human Gene 1.0 ST Array 8176790 - -
          Affymetrix Human Gene 1.0 ST Array 8176793 - -
          Affymetrix Human Gene 1.0 ST Array 8176791 - -
          Affymetrix Human Gene 1.0 ST Array 8176792 - -
          Affymetrix Human Gene 1.1 ST Array 8176789 - -
          Affymetrix HT HG-U133+ PM Array Plate 216544_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1552952_PM_at - -
          Human Genome U133 Plus 2.0 Array 1552952_PM_at - -
          Human Genome U133 Plus 2.0 Array 1552952_at - -
          Human Genome U133 Plus 2.0 Array 216544_PM_at - -
          Human Genome U133 Plus 2.0 Array 216544_at - -
          Human Genome U133A 2.0 Array 216544_at - -
          Human Genome U133A 2.0 Array 216544_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NR_002193_at - -
          AffymetrixPathwork Diagnostics Pathchip pw525977 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216544_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216544_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216544_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216544_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552952_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216544_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552952_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552952_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552952_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216544_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216544_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216544_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216544_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552952_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552952_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216544_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552952_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552952_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216544_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552952_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552952_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552952_at5 - -
          Human Genome U133 Set 216544_at - -
          Human Genome U133 Set 216544_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 227467 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2145781 BC026077.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2145781 NR_002193.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1654284 - -
          HumanHT-12 V4.0 expression beadchip ILMN_2145781 BC026077.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2145781 NR_002193.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2145781 BC026077.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2145781 NR_002193.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2145781 BC026077.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2145781 NR_002193.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1654284 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 6450193 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_15270 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2145781 BC026077.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2145781 NR_002193.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2145781 BC026077.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2145781 NR_002193.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2145781 BC026077.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2145781 NR_002193.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1654284 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2145781 BC026077.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2145781 NR_002193.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline