Search
Search

LINC00964| Gene Symbol: LINC00964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC033393.1 | - | 1 | 1744 | 97 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105375742 | LOC105375742 | Sense | Chr. 8 - 124936331 - 124941533 on NCBI Build GRCh38 |
Set Membership: |
|