Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03668510_s1
          See other LINC01605 GE Assays ›
          Gene Symbol:
          LINC01605
          Gene Name:
          long intergenic non-protein coding RNA 1605
          Gene Aliases:
          LincDUSP, ODIR1, lncGALM
          Chromosome Location:
          Chr.8: 37403516 - 37599839 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03668510_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01605
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA OA989052.1 - 100
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB341617.1 - Chr. 8 - 37597485 - 37598011 [+] on Build GRCh38 100
          GenBankmRNA AI809548.1 - Chr. 8 - 37597516 - 37598030 [+] on Build GRCh38 100
          GenBankmRNA BF001923.1 - Chr. 8 - 37597487 - 37597821 [+] on Build GRCh38 100
          GenBankmRNA BX103959.1 - Chr. 8 - 37597497 - 37598169 [-] on Build GRCh38 100

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Homo sapiens

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357349 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075466 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075420 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075445 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075439 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075417 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16087184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075462 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357352 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357459 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075475 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16087175 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075470 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357449 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075442 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075513 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075518 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075451 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075405 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357389 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357376 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075435 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075502 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075450 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075437 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075468 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075441 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075433 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075452 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075488 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075494 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075465 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075472 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075485 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075406 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357344 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357417 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357416 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075478 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357388 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075423 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075408 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357378 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075486 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075449 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16087189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075469 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16087182 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075458 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357382 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357443 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075489 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075480 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075440 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357371 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075487 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075453 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075430 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075463 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357392 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357405 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075459 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075506 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075404 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075497 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075483 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16087180 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075427 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075476 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075416 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357361 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075438 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075474 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357348 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357445 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075413 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075429 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357363 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26357377 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06075407 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 176916 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 461801 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3802116 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P751220 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 32178 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 15277 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3802116 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P751220 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P751220 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 177216 AK128026.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 177216 BC132719.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 177216 NM_194300.2 refseq_mrna_human_NM
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01899677 - -
          Agilent Human 1B Microarray G4111A A_32_P341516 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 232040_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010127.hg.1 ENST00000522471.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010127.hg.1 ENST00000522471.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010127.hg.1 ENST00000519691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010127.hg.1 ENST00000522471.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010127.hg.1 ENST00000522471.1 Ensembl
          Human Genome U133 Plus 2.0 Array 232040_PM_at - -
          Human Genome U133 Plus 2.0 Array 232040_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232040_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232040_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232040_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232040_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232040_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232040_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232040_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232040_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232040_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232040_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232040_at1 - -
          Human Genome U95 Set 59497_PM_g_at - -
          Human Genome U95 Set 59495_PM_at - -
          Human Genome U95 Set 59495_at - -
          Human Genome U95 Set 59497_g_at - -
          Human Genome U95 Set 84580_PM_at - -
          Human Genome U95 Set 84580_at - -
          Human Genome U133 Set 232040_PM_at - -
          Human Genome U133 Set 232040_at - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1822338 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_80188 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_80188 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1835847 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1835847 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1822338 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline